Learn More
AURORA BIOLABS LLC Human Malic enzyme 3 (ME3),100 μg
Catalog No. NC3787984
Encompass
ME3 is another isoform of malic enzyme that is found in various tissues in the human body. This gene encodes a mitochondrial NADP(+)- dependent isoform. Multiple alternatively spliced transcript variants have been found for this gene, but the biological validity of some variants has not been determined. Its exact biochemical significance may vary depending on the tissue or cell type in which it is expressed.
Catalog No. NC3787984 Supplier AURORA BIOLABS LLC Supplier No. 180003100
ME3 is another isoform of malic enzyme that is found in various tissues in the human body. This gene encodes a mitochondrial NADP(+)- dependent isoform. Multiple alternatively spliced transcript variants have been found for this gene, but the biological validity of some variants has not been determined. Its exact biochemical significance may vary depending on the tissue or cell type in which it is expressed.