Learn More
American Research Products Inc Human Mammaglobin A SCGB2A2
Catalog No. 5000328420
Encompass
Human Mammaglobin-A (SCGB2A2) ELISA kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28420 Supplier American Research Products Inc Supplier No. CSBEL020817HU
Human Mammaglobin-A (SCGB2A2) ELISA kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich