Learn More
American Research Products Inc Human mammalian target of rapa
Catalog No. 5000329509
Encompass
Human mammalian target of rapamyein mTOR ELISA Kit from CUSABIO Detection Range 1 56 pg/mL-100 pg/mL Sensitivity 0 39 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29509 Supplier American Research Products Inc Supplier No. CSBE09038H
Human mammalian target of rapamyein mTOR ELISA Kit from CUSABIO Detection Range 1 56 pg/mL-100 pg/mL Sensitivity 0 39 pg/mL Sample serum plasma tissue homogenates Method Sandwich