Learn More
Biomatik Corporation Human Mannosidase Alpha Class 1A Member 1 (MAN1A1) ELISA Kit
Supplier:  Biomatik Corporation EKU05800
Encompass_Preferred
Human Mannosidase Alpha Class 1A Member 1 (MAN1A1) ELISA Kit for detecting MAN1A1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P33908. Gene ID: 4121. Target Synonyms: HUMM3; HUMM9; MAN9; Mannosyl-Oligosaccharide 1,2-Alpha-Mannosidase IA; Mannosidase alpha class 1A member 1; Processing alpha-1,2-mannosidase IA. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4872