Learn More
ACROBiosystems Human MAOB, His Tag (MAB-H5547) is expressed from Baculovirus-Insect cells. It contains AA Ser 2 - Val 520 (Accession # P27338-1).
Supplier:  ACROBiosystems MABH5547500UG
Encompass_Preferred
Monoamine Oxidase Type B (MAOB) is an enzyme located in the outer mitochondrial membrane. It catalyzes the oxidative deamination of biogenic and xenobiotic amines and plays an important role in the breakdown of dopamine in the brain. MAOB inhibitors, that reduces dopamine turnover, are in clinical use for the treatment of Parkinson's disease (PD).
Catalog No. 50-225-3871