Learn More
Sino Biological Recombinant Human p38 Protein (His Tag), Activated in vitro, HPLC-verified
Supplier:  Sino Biological 10081H07BA20
Encompass_Preferred
The amino acids cprresponding to the full length of human MAPK14 isoform 1 (NP_001306.1) (Met1-Ser360) was fused with a polyhistidine tag at the N-terminus. Activated in vitro by MAP2K6 (10422-H20B1).
Catalog No. 50-255-5503