Learn More
Sino Biological Anti-JNK2 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10745MM06100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human JNK2 / MAPK9 (rh JNK2 / MAPK9; Catalog#10745-H08B; NP_002743.3; Met1-Arg424). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-1218