Learn More
Sino Biological Anti-JNK2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10745RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human JNK2 / MAPK9 (rh JNK2 / MAPK9; Catalog#10745-H08B; NP_002743.3; Met 1-Arg 424). JNK2 / MAPK9 specific IgG was purified by Human JNK2 / MAPK9 affinity chromatography.
Catalog No. 50-240-9050