Learn More
Sino Biological Human MAPT PAb 100ul (Sino Biological)
Supplier:  Sino Biological 10058T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human MAPT / PHF-tau (rh MAPT / PHF-tau; Catalog#10058-H07E; NP_058525.1; Ala2-Leu352). MAPT / PHF-tau specific IgG was purified by Human MAPT / PHF-tau affinity chromatography.
Catalog No. 50-237-1415