Learn More
Sino Biological Human Tau / TAU412 Protein (isoform 1N4R, His Tag)
Supplier:  Sino Biological 29992H07E100
Encompass_Preferred
A DNA sequence encoding the Human Tau / TAU412 (NP_001116539.1) (Ala2-Leu412) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-255-3814