Learn More
Sino Biological Human Tau / TAU410 Protein (isoform 2N3R, His Tag)
Supplier:  Sino Biological 30133H07E100
Encompass_Preferred
A DNA sequence encoding the Human MAPT (P10636-5) (Ala2-Leu410) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-255-3803