Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human MARCH10 (aa 379-475) Control Fragment Recombinant Protein

Catalog No. RP93424
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP93424 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP93424 Supplier Invitrogen™ Supplier No. RP93424
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (32%), Rat (32%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-54763. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

MARCH10 is a member of the MARCH family of membrane-bound E3 ubiquitin ligases to target lysines in substrate proteins, thereby signaling their vesicular transport between membrane compartments.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8NA82
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 162333
Name Human MARCH10 (aa 379-475) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 10-Mar; 4933417C16Rik; AV039223; FLJ35757; March10; MARCH-X; membrane associated ring finger 10; membrane associated ring-CH-type finger 10; membrane-associated ring finger (C3HC4) 10; membrane-associated ring finger (C3HC4) 10, E3 ubiquitin protein ligase; membrane-associated RING finger protein 10; Membrane-associated RING-CH protein X; microtubule-associated E3 ubiquitin ligase; OTTMUSG00000002847; Probable E3 ubiquitin-protein ligase MARCH10; RGD1311692; RING finger protein 190; RING-type E3 ubiquitin transferase MARCH10; Rnf190; testis secretory sperm-binding protein Li 228n
Common Name MARCH10
Gene Symbol MARCH10
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LPDRESATEKDRGGSENAKKSPLSWDTKSEPRQEVGVNAENVWSDCISVEHRPGTHDSEGYWKDYLNSSQNSLDYFISGRPISPRSSVNSSYNPPAS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.