Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human MARK1 Control Fragment Recombinant Protein

Catalog No. RP89777
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP89777 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP89777 Supplier Invitrogen™ Supplier No. RP89777
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (93%), Rat (93%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-110740. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

MARK1 (MARK) is a serine/threonine kinase which is involved in cytoskeletal organization and biogenesis. Involved in cell polarity by phosphorylating the microtubule-associated proteins MAP2, MAP4 and MAPT/TAU at KXGS motifs, causing detachment from microtubules, and their disassembly. MARK1 is involved in the regulation of neuronal migration through its dual activities in regulating cellular polarity and microtubule dynamics, possibly by phosphorylating and regulating DCX. Also acts as a positive regulator of the Wnt signaling pathway, probably by mediating phosphorylation of dishevelled proteins (DVL1, DVL2 and/or DVL3).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9P0L2
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 4139
Name Human MARK1 Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias AW491150; B930025N23Rik; ELKL motif serine/threonine-protein kinase 3; ELKL motif serine-threonine protein kinase 3; Emk3; KIAA1477; MAP/microtubule affinity regulating kinase 1; MAP/microtubule affinity-regulating kinase 1; MARK; MARK1; mark1 {ECO:0000312; microtubule affinity regulating kinase 1; mKIAA1477; mPar-1c; PAR1 homolog c; Par1c; Par-1c; RGD:619882}; serine/threonine-protein kinase MARK1
Common Name MARK1
Gene Symbol MARK1
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence QEEEELKPYTEPDPDFNDTKRIDIMVTMGFARDEINDALINQKYDEVMATYILLGRKPPEFEGGESLSSGNLCQRSRPSSDLNNSTLQSPAHLKVQRSISANQKQRRFSDHAGPSIPPAVSYTKRPQANSVESEQK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.