Learn More
Biomatik Corporation Human MAS Related GPR Member X2 (MRGPRX2) ELISA Kit
Supplier:  Biomatik Corporation EKU12096
Encompass_Preferred
Human MAS Related GPR Member X2 (MRGPRX2) ELISA Kit for detecting MRGPRX2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q96LB1. Gene ID: 117194. Target Synonyms: MRGX2; Mas-related G-protein coupled receptor member X2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8031