Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human MASP1 Full-length ORF (NP_001027019.1, 1 a.a. - 380 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89951893 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-951-893 25 ug
89-951-892 10 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-951-893 Supplier Abnova™ Supplier No. H00005648P01L
Only null left
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The Ra-reactive factor (RARF) is a complement-dependent bactericidal factor that binds to the Ra and R2 polysaccharides expressed by certain enterobacteria. Alternate splicing of this gene results in multiple transcript variants encoding two RARF components that are involved in the mannan-binding lectin (MBL) pathway of complement activation. Two of the isoforms are cleaved into two chains which form a heterodimer linked by a disulfide bond. The encoded proteins are members of the trypsin family of peptidases. [provided by RefSeq]

Sequence: MRWLLLYYALCFSLSKASAHTVELNNMFGQIQSPGYPDSYPSDSEVTWNITVPDGFRIKLYFMHFNLESSYLCEYDYVKVETEDQVLATFCGRETTDTEQTPGQEVVLSPGSFMSITFRSDFSNEERFTGFDAHYMAVDVDECKEREDEELSCDHYCHNYIGGYYCSCRFGYILHTDNRTCRVECSDNLFTQRTGVITSPDFPNPYPKSSECLYTIELEEGFMVNLQFEDIFDIEDHPEVPCPYDYIKIKVGPKVLGPFCGEKAPEPISTQSHSVLILFHSDNSGENRGWRLSYRAAGNECPELQPPVHGKIEPSQAKYFFKDQVLVSCDTGYKVLKDNVEMDTFQIECLKDGTWSNKIPTCKKNEIDLESELKSEQVTE

Specifications

Accession Number NP_001027019.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 5648
Molecular Weight (g/mol) 70kDa
Name MASP1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 ug
Immunogen MRWLLLYYALCFSLSKASAHTVELNNMFGQIQSPGYPDSYPSDSEVTWNITVPDGFRIKLYFMHFNLESSYLCEYDYVKVETEDQVLATFCGRETTDTEQTPGQEVVLSPGSFMSITFRSDFSNEERFTGFDAHYMAVDVDECKEREDEELSCDHYCHNYIGGYYCSCRFGYILHTDNRTCRVECSDNLFTQRTGVITSPDFPNPYPKSSECLYTIELEEGFMVNLQFEDIFDIEDHPEVPCPYDYIKIKVGPKVLGPFCGEKAPEPISTQSHSVLILFHSDNSGENRGWRLSYRAAGNECPELQPPVHGKIEPSQAKYFFKDQVLVSCDTGYKVLKDNVEMDTFQIECLKDGTWSNKIPTCKKNEIDLESELKSEQVTE
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CRARF/CRARF1/DKFZp686I01199/FLJ26383/MASP/MGC126283/MGC126284/PRSS5/RaRF
Common Name MASP1
Gene Symbol MASP1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.