Learn More
American Research Products Inc Human Mast Cell CarboXypeptida
Catalog No. 5000329330
Encompass
Human Mast Cell CarboXypeptidase ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29330 Supplier American Research Products Inc Supplier No. CSBE13756H
Human Mast Cell CarboXypeptidase ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich