Learn More
ATCC Human mastadenovirus B7; Strain: Gomen
Catalog No. 502384239
Encompass
Human adenovirus 7 strain Gomen is propagated in HeLa cells (ATCC CCL-2). This strain was isolated in 1954 from the throat washing of a military recruit with pharyngitis in California. The strain has applications in respiratory disease research.
Catalog No. 50-238-4239 Supplier ATCC Supplier No. VR7
Human adenovirus 7 strain Gomen is propagated in HeLa cells (ATCC CCL-2). This strain was isolated in 1954 from the throat washing of a military recruit with pharyngitis in California. The strain has applications in respiratory disease research.