Learn More
Sino Biological Human mastadenovirus E (HAdV-E) fiber Antibody, Rabbit PAb, Antigen Affinity Purified 20 µL
Catalog No. 5000749259
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human mastadenovirus E (HAdV-E) fiber ( Catalog#40625-V08B; YP_068048.1; Met1-Gln425). Human mastadenovirus E (HAdV-E) fiber specific IgG was purified by Human mastadenovirus E (HAdV-E) fiber affinity chromatography.
Catalog No. 50-007-49259 Supplier Sino Biological Supplier No. 40625T6220
Produced in rabbits immunized with purified, recombinant Human mastadenovirus E (HAdV-E) fiber ( Catalog#40625-V08B; YP_068048.1; Met1-Gln425). Human mastadenovirus E (HAdV-E) fiber specific IgG was purified by Human mastadenovirus E (HAdV-E) fiber affinity chromatography.