Learn More
ATCC Human adenovirus 40 Strain: Dugan
Catalog No. 502383287
Encompass
Human adenovirus 40 strain Dugan [79-18025] is propagated in 293 [HEK-293] cells (ATCC CRL-1573). This strain was isolated in 1979 from the feces of a human with infantile gastroenteritis. This product is whole-genome sequenced has applications in enteric and infectious disease research.
Catalog No. 50-238-3287 Supplier ATCC Supplier No. VR931
Human adenovirus 40 strain Dugan [79-18025] is propagated in 293 [HEK-293] cells (ATCC CRL-1573). This strain was isolated in 1979 from the feces of a human with infantile gastroenteritis. This product is whole-genome sequenced has applications in enteric and infectious disease research.