Learn More
Zymo Research Corporation Human Matched DNA Set
SDP
Catalog No. 502037112
Encompass_Preferred
This set is an ideal control for detection and/or quantification methods against 5- methylcytosine (5mC) and 5-hydroxymethylcytosine (5hmC) as both modified cytosines are present at physiologically relevant levels and loci.
Catalog No. 50-203-7112 Supplier Zymo Research Corporation Supplier No. D5018
This set is an ideal control for detection and/or quantification methods against 5- methylcytosine (5mC) and 5-hydroxymethylcytosine (5hmC) as both modified cytosines are present at physiologically relevant levels and loci.