Learn More
ACROBiosystems Human Matriptase / ST14 Catalytic Domain Protein, His Tag
Catalog No. 502329115
Encompass_Preferred
Human Matriptase / ST14 Catalytic Domain Protein, His Tag (MAE-H5143) is expressed from E.coli cells. It contains AA Gly 596 - Val 855 (Accession # NP_068813).
Catalog No. 50-232-9115 Supplier ACROBiosystems Supplier No. MAEH51431MG
Human Matriptase / ST14 Catalytic Domain Protein, His Tag (MAE-H5143) is expressed from E.coli cells. It contains AA Gly 596 - Val 855 (Accession # NP_068813).