Learn More
Bioworld HUMAN MATRIX METALLOPROTEIN
Supplier:  Bioworld 220604141
Encompass_Preferred
HUMAN MATRIX METALLOPROTEINASE-13 (RECOMBINANT): Recombinant Human Matrix Metalloproteinase-13 (MMP-13) Source:E.Coli Background: Description: Latent recombinant human pro-collagenase (MMP-13) also called collagenase-3 truncated fr...
Catalog No. 50-253-510