Learn More
Biomatik Corporation Human Matrix Metalloproteinase 8 (MMP8) CLIA Kit
Supplier:  Biomatik Corporation EKU09448
Encompass_Preferred
Human Matrix Metalloproteinase 8 (MMP8) CLIA Kit for detecting MMP8 in serum, plasma, tissue homogenates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.43-2,500pg/ml. Sensitivity: 1.34pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P22894. Gene ID: 4317. Target Synonyms: CLG2; HNC; PMNL-CL; Collagenase 2; Neutrophil Collagenase; PMNL collagenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5627