Learn More
Biomatik Corporation Human Matrix Metalloproteinase 9 (MMP9) CLIA Kit
Supplier:  Biomatik Corporation EKU08891
Encompass_Preferred
Human Matrix Metalloproteinase 9 (MMP9) CLIA Kit for detecting MMP9 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 41.2-30,000pg/ml. Sensitivity: 15.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P14780. Gene ID: 4318. Target Synonyms: GELB; Gelatinase B; CLG4B; CLG4-B; 92 KDa Gelatinase; 92kDa Type IV Collagenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8601