Learn More
Biomatik Corporation Human Matrix Metalloproteinase 23B (MMP23B) ELISA Kit
Supplier:  Biomatik Corporation EKU05855
Encompass_Preferred
Human Matrix Metalloproteinase 23B (MMP23B) ELISA Kit for detecting MMP23B in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5,000pg/ml. Sensitivity: 31pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O75900. Gene ID: 8510. Target Synonyms: MMP22; MMP23-B; MIFR; MIFR-1; Matrix Metalloproteinase 22; Femalysin; Matrix Metalloproteinase In The Female Reproductive Tract. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8880