Learn More
Biomatik Corporation Human Matrix Metalloproteinase 10 (MMP10) CLIA Kit
Supplier:  Biomatik Corporation EKU09563
Encompass_Preferred
Human Matrix Metalloproteinase 10 (MMP10) CLIA Kit for detecting MMP10 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 6.86-5,000pg/ml. Sensitivity: 2.72pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P09238. Gene ID: 4319. Target Synonyms: STMY2; SL2; Stromelysin 2; Transin-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5335