Learn More
Sino Biological Human MAX PAb 100ul (Sino Biological)
Supplier:  Sino Biological 12885T52100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human MYC associated factor X (rh MYC associated factor X; Catalog#12885-H20B; NP_002373; Met1-Ser160). MYC associated factor X specific IgG was purified by Human MYC associated factor X affinity chromatography.
Catalog No. 50-237-1461