Learn More
Sino Biological Human MBD3 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 11692T6220
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human MBD3 isoform 2 (O95983-2; Met 1-Val 259). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-237-4257