Learn More
Sino Biological Anti-Mannan Binding Lectin/MBL2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 201183T08100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human Mannan Binding Lectin / MBL2 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-241-1744