Learn More
Sino Biological Recombinant Human CD146/MCAM Protein (His Tag)
Supplier:  Sino Biological 10115H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human CD146 precursor (NP_006491.2) (Met 1-Gly 559) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-255-5467