Learn More
Sino Biological Recombinant Human CD146/MCAM Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 10115H08HB100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human CD146 precursor (NP_006491.2) (Met1-Gly559) was expressed with a C-terminal polyhistidine tag. The purified protein was biotinylated in vitro.
Catalog No. 50-255-5466