Learn More
FUJIFILM BIOSCIENCES INC Human MCP-1 (CCL2), Recombinant Protein, Shenandoah Biotechnology, 5 ug vial
Supplier:  FUJIFILM BIOSCIENCES INC 100135UG
Encompass_Preferred
Monocyte chemotactic protein 1 (MCP-1), also known as CCL2, is produced by injured or infected tissues. MCP-1 signals through the CCR2 and CCR4 G protein-coupled receptors to recruit memory T cells, monocytes, and dendritic cells to sites of inflammation.
Catalog No. 50-990-650