Learn More
FUJIFILM Biosciences Human MCP-2 (CCL8), Recombinant Protein, Shenandoah Biotechnology, 10 ug vial
Catalog No. 50900486
Encompass_Preferred
Monocyte chemotactic protein 2 (MCP-2), also known as CCL8, is a cytokine that is important during allergic and inflammatory responses. MCP-2 activates mast cells, eosinophils, and basophils through the G protein-coupled chemokine receptors CCR1, CCR2B, and CCR5. MCP-2 signaling through the CCR5 receptor also functions as a natural inhibitor of the human immunodeficiency virus, type 1 (HIV-1).
Catalog No. 50-900-486 Supplier FUJIFILM Biosciences Supplier No. 10013010UG
Monocyte chemotactic protein 2 (MCP-2), also known as CCL8, is a cytokine that is important during allergic and inflammatory responses. MCP-2 activates mast cells, eosinophils, and basophils through the G protein-coupled chemokine receptors CCR1, CCR2B, and CCR5. MCP-2 signaling through the CCR5 receptor also functions as a natural inhibitor of the human immunodeficiency virus, type 1 (HIV-1).