Learn More
FUJIFILM Biosciences Human MCP-3 (CCL7), Recombinant Protein, Shenandoah Biotechnology, 100 ug vial
Catalog No. 50990656
Encompass_Preferred
Monocyte chemotactic protein 3 (MCP-3), also called CCL7, is produced by macrophages and tumor cell lines. MCP-3 signals through the G protein-coupled receptors CCR1, CCR2, and CCR3. MCP-3 chemoattracts monocytes and regulates macrophage function during inflammation and metastasis.
Catalog No. 50-990-656 Supplier FUJIFILM Biosciences Supplier No. 100131100UG
Monocyte chemotactic protein 3 (MCP-3), also called CCL7, is produced by macrophages and tumor cell lines. MCP-3 signals through the G protein-coupled receptors CCR1, CCR2, and CCR3. MCP-3 chemoattracts monocytes and regulates macrophage function during inflammation and metastasis.