Learn More
FUJIFILM BIOSCIENCES INC Human MCP-3 (CCL7), Recombinant Protein, Shenandoah Biotechnology, 10 ug vial
Supplier:  FUJIFILM BIOSCIENCES INC 10013110UG
Encompass_Preferred
Monocyte chemotactic protein 3 (MCP-3), also called CCL7, is produced by macrophages and tumor cell lines. MCP-3 signals through the G protein-coupled receptors CCR1, CCR2, and CCR3. MCP-3 chemoattracts monocytes and regulates macrophage function during inflammation and metastasis.
Catalog No. 50-990-655