Learn More
FUJIFILM Biosciences Human MCP-4 (CCL13), Recombinant Protein, Shenandoah Biotechnology, 20 ug vial
Catalog No. 50990659
Encompass_Preferred
Monocyte chemotactic protein 4 (MCP-4), also called CCL13, is induced by inflammatory proteins such as interleukin 1 (IL-1) and tumor necrosis factor alpha (TNF-A). MCP-4 is a ligand for the G protein coupled chemokine receptors CCR2, CCR3, and CCR5. MCP-4 activates signaling in monocytes, T lymphocytes, eosinophils, and basophils during inflammation and allergic responses.
Catalog No. 50-990-659 Supplier FUJIFILM Biosciences Supplier No. 10013220UG
Monocyte chemotactic protein 4 (MCP-4), also called CCL13, is induced by inflammatory proteins such as interleukin 1 (IL-1) and tumor necrosis factor alpha (TNF-A). MCP-4 is a ligand for the G protein coupled chemokine receptors CCR2, CCR3, and CCR5. MCP-4 activates signaling in monocytes, T lymphocytes, eosinophils, and basophils during inflammation and allergic responses.