Learn More
FUJIFILM BIOSCIENCES INC Human MCP-4 (CCL13), Recombinant Protein, Shenandoah Biotechnology, 5 ug vial
Supplier:  FUJIFILM BIOSCIENCES INC 1001325UG
Encompass_Preferred
Monocyte chemotactic protein 4 (MCP-4), also called CCL13, is induced by inflammatory proteins such as interleukin 1 (IL-1) and tumor necrosis factor alpha (TNF-A). MCP-4 is a ligand for the G protein coupled chemokine receptors CCR2, CCR3, and CCR5. MCP-4 activates signaling in monocytes, T lymphocytes, eosinophils, and basophils during inflammation and allergic responses.
Catalog No. 50-990-658