Learn More
Abcam Human MCP1 Matched Antibody Pair Kit, 5 x 96 Tests
Catalog No. 502657549
Encompass_Preferred
Human MCP1 Matched Antibody Pair Kit is a kit for ELISA development for the measurement of Human MCP1 by sandwich ELISA method.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-7549 Supplier Abcam Supplier No. AB2144765X96TESTS
Human MCP1 Matched Antibody Pair Kit is a kit for ELISA development for the measurement of Human MCP1 by sandwich ELISA method.
The product is subject to the following: Abcam Restricted Use Statement