Learn More
BIOHIPPO Human MCSF Microsample
SDP
Catalog No. 5000315519
Encompass
Human MCSF Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MCSF in serum plasma urine tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 7 82-500 pg/mL with a sensitivity of 3 1 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human MCSF Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human MCSF Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human MCSF biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15519 Supplier BIOHIPPO Supplier No. BHE15212304
Human MCSF Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MCSF in serum plasma urine tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 7 82-500 pg/mL with a sensitivity of 3 1 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human MCSF Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human MCSF Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human MCSF biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color