Learn More
BIOHIPPO Human MCSFR Microsample
SDP
Catalog No. 5000315106
Encompass
Human MCSFR Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MCSFR in tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 127 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human MCSFR Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human MCSFR Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human MCSFR biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15106 Supplier BIOHIPPO Supplier No. BHE15211335
Human MCSFR Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MCSFR in tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 127 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human MCSFR Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human MCSFR Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human MCSFR biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color