Learn More
Raybiotech Inc Human MD-2 (LY96) ELISA Kit
Supplier:  Raybiotech Inc ELHMD21
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 1.6 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes human MD-2. Other species not yet determined.
Catalog No. 50-194-9532