Learn More
Abcam Human MDH1 ELISA Kit (cytosolic malate dehydrogenase), 1 x 96 Tests
Catalog No. 502663206
Encompass_Preferred
Human MDH1 ELISA Kit (cytosolic malate dehydrogenase) is a single-wash 90-min Simplestep used to quantify Human MDH1 (cytosolic malate dehydrogenase) with a sensitivity of 64 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-266-3206 Supplier Abcam Supplier No. AB2557241X96TESTS
Add to Cart
Add to Cart
Human MDH1 ELISA Kit (cytosolic malate dehydrogenase) is a single-wash 90-min Simplestep used to quantify Human MDH1 (cytosolic malate dehydrogenase) with a sensitivity of 64 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement