Learn More
Raybiotech Inc Human Melan-A/MART-1 ELISA Kit
Supplier:  Raybiotech Inc ELHMLANA1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.045 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human Melan-A.
Catalog No. 50-194-9553