Learn More
American Research Products Inc Human melanin concentrating ho
Catalog No. 5000327230
Encompass
Human melanin-concentrating hormone (PMCH) ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27230 Supplier American Research Products Inc Supplier No. CSBEL018230HU
Human melanin-concentrating hormone (PMCH) ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich