Learn More
Biomatik Corporation Human Melanocortin 5 Receptor (MC5R) CLIA Kit
Supplier:  Biomatik Corporation EKU10014
Encompass_Preferred
Human Melanocortin 5 Receptor (MC5R) CLIA Kit for detecting MC5R in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 13.7-10,000pg/ml. Sensitivity: 5.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P33032. Gene ID: 4161. Target Synonyms: MC-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5866