Learn More
ACROBiosystems Human Melanotransferrin, His Tag (MEN-H5223) is expressed from human 293 cells (HEK293). It contains AA Asp 358 - Ser 706 (Accession # P08582-1).
Catalog No. 502254034
Encompass_Preferred
Melanotransferrin (MTF), also known as CD228, P97, MELTF and MFI2, is a cell-surface glycoprotein found on melanoma cells. The protein shares sequence similarity and iron-binding properties with members of the transferrin superfamily. Involved in iron cellular uptake. Seems to be internalized and then recycled back to the cell membrane. Binds a single atom of iron per subunit. Could also bind zinc. MTF expression is low in normal tissues but high in tumor and embryonic tissues. In melanoma MTF is associated with tumor metastasis and angiogenesis and upregulated in lung cancer tissue and cell lines. In the brain MTF is expressed in capillary endothelium but also in the reactive microglia associated with senile plaques in Alzheimer's disease (AD).
This item is not returnable. View return policy
Catalog No. 50-225-4034 Supplier ACROBiosystems Supplier No. MENH5223100UG
Melanotransferrin (MTF), also known as CD228, P97, MELTF and MFI2, is a cell-surface glycoprotein found on melanoma cells. The protein shares sequence similarity and iron-binding properties with members of the transferrin superfamily. Involved in iron cellular uptake. Seems to be internalized and then recycled back to the cell membrane. Binds a single atom of iron per subunit. Could also bind zinc. MTF expression is low in normal tissues but high in tumor and embryonic tissues. In melanoma MTF is associated with tumor metastasis and angiogenesis and upregulated in lung cancer tissue and cell lines. In the brain MTF is expressed in capillary endothelium but also in the reactive microglia associated with senile plaques in Alzheimer's disease (AD).