Learn More
BIORTUS USA INC Human MELK Protein, 100UG
Supplier:  BIORTUS USA INC BP0626901A
Encompass_Preferred
Gene Name: MELK. Uniprot: Q14680. Construct: 6His-TEV-MELK(D3-V330). Expression Host: E. coli. Purity: 90%.
Catalog No. NC2953417