Learn More
Sino Biological Anti-Melanotransferrin/MFI2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 200227T32100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human MFI2 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-241-1199