Learn More
Biomatik Corporation Human Membrane Protein, Palmitoylated 6 (MPP6) ELISA Kit
Supplier:  Biomatik Corporation EKU05905
Encompass_Preferred
Human Membrane Protein, Palmitoylated 6 (MPP6) ELISA Kit for detecting MPP6 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 12.5-800pg/ml. Sensitivity: 4.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9NZW5. Gene ID: 51678. Target Synonyms: PALS2; VAM1; P55T; MAGUK P55 Subfamily Member 6; Veli-associated MAGUK 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6520