Learn More
STEMCELL Technologies EasySep™ Human Memory CD4+ T Cell Enrichment Kit, For processing: 1 x 10^9 cells
SDP
Catalog No. NC0651016
Encompass
The EasySep™ Human Memory CD4 T Cell Enrichment Kit is designed to isolate memory CD4+ T cells from fresh or previously frozen peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD8, CD14, CD16, CD19, CD20, CD36, CD45RA, CD56, CD123, TCRγ/δ, glycophorin A and dextran-coated magnetic particles. The labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.
Catalog No. NC0651016 Supplier STEMCELL Technologies Supplier No. 19157
The EasySep™ Human Memory CD4 T Cell Enrichment Kit is designed to isolate memory CD4+ T cells from fresh or previously frozen peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD8, CD14, CD16, CD19, CD20, CD36, CD45RA, CD56, CD123, TCRγ/δ, glycophorin A and dextran-coated magnetic particles. The labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.